missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol 99.5+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 500 mL |
| Physical Form | Clear Liquid at 20°C |
| UN Number | 1219 |
| Boiling Point | 82°C |
| Formula Weight | 60.10 |
| Percent Purity | ≥99.5% (GC) |
| Chemical Name or Material | Isopropyl Alcohol [for HPLC Solvent] |
Safety and Handling
| EINECSNumber | (2)-0207 |
| RTECSNumber | NT8050000 |
| TSCA | Yes |