missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, Exceeds A.C.S. Specifications, HPLC Grade, 99.7%, Spectrum™ Chemical

Catalog No. p-7157584
Change view
Click to view available options
Quantity:
4 L
4 x 4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603538 4 L Amber Glass Bottle
18603540 4 x 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18603538 Supplier Spectrum Chemical Mfg Cor Supplier No. HP6924LT
Only null left

HP692, 67-63-0, C3H8O

Spectrum™ Chemical Isopropyl Alcohol, Exceeds A.C.S. Specifications, HPLC Grade solvents are lot-tested and precisely controlled to meet the high quality requirements of chromatographic analysis including high purity, low UV-absorbance, guaranteed suitability for fluorescence detection, low particulates (eliminates pre-filtration) and controlled low-content of non-volatile components. All Isopropyl Alcohol chromatography solvents are delivered with an expiration date.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 100%
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation 2 ppm
UN Number UN1219
Water 0.05%
Boiling Point 78.3°C
Percent Purity 99.7%
Grade High Purity
Chemical Name or Material Isopropyl Alcohol