missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, Exceeds A.C.S. Specifications, HPLC Grade, 99.7%, Spectrum™ Chemical
Description
Spectrum™ Chemical Isopropyl Alcohol, Exceeds A.C.S. Specifications, HPLC Grade solvents are lot-tested and precisely controlled to meet the high quality requirements of chromatographic analysis including high purity, low UV-absorbance, guaranteed suitability for fluorescence detection, low particulates (eliminates pre-filtration) and controlled low-content of non-volatile components. All Isopropyl Alcohol chromatography solvents are delivered with an expiration date.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 4 x 4 L |
| Packaging | Amber Glass Bottle |
| Residue after Evaporation | 2 ppm |
| UN Number | UN1219 |
| Water | 0.05% |
| Boiling Point | 78.3°C |
| Percent Purity | 99.7% |
| Grade | High Purity |
| Chemical Name or Material | Isopropyl Alcohol |