missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, Certified, 70% (v/v), LabChem™

Supplier:  LabChem LC15760200

 View more versions of this product

Catalog No. LC15760200

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Isopropyl Alcohol
67-63-0,7732-18-5
70,30
0.88g/mL
200 L
Passes Test
Liquid
C3H8O
2.5316mm2/s
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.09
Certified
-88°C
Colorless
Carbon dioxide; Carbon monoxide
82°C
12°C
Poly Drum
0.88g/mL +/- 0.005
C3H8O
UN1219
Soluble in water
CC(C)O
60.096
CHEBI:17824
70% (v/v)

Chemical Identifiers

67-63-0
60.096
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.

Danger

EINECSNumber : 200-661-7

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions