missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, Certified, 50% (v/v), LabChem™

$163.32 - $585.71

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
LC157554
View Documents
LabChem
LC157554
4 L Poly Bottle
Each for $163.32
Only null left
 
LC157555
View Documents
LabChem
LC157555
20 L Cubitainer
Each for $585.71
Only null left
 
Description

Description

Specifications

Chemical Identifiers

67-63-0
60.096
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol

Specifications

-88°C
Colorless
Carbon dioxide; Carbon monoxide
82°C
12°C
Poly Bottle
0.921g/mL ±0.006
C3H8O
UN1219
Soluble in water
CC(C)O
60.096
CHEBI:17824
50% (v/v)
Isopropyl Alcohol
67-63-0,7732-18-5
50,50
0.92g/mL
4 L
Passes Test
Liquid
C3H8O
2.5316mm2/s
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.09
Certified
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing mist, spray, vapors.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a poison center/doctor if you feel unwell.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

EINECSNumber : 200-661-7

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'