missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, B&J Brand™ LC-MS, for LC-MS and HPLC, >99.9%, Honeywell Burdick & Jackson

Catalog No. p-7112034
Change view
Click to view available options
Quantity:
4 x 2.5 L
4 x 4 L
6 x 1 L
Packaging:
GL45 Glass Bottle
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000141 6 x 1 L GL45 Glass Bottle
6000139 4 x 2.5 L GL45 Glass Bottle
6000140 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6000141 Supplier Honeywell Chemicals Supplier No. LC3231
Only null left

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 100.0000%
Density 0.785 g/cm3
Boiling Point 82.3°C
Quantity 6 x 1 L
Flash Point 12°C
Packaging GL45 Glass Bottle
UN Number UN1219
Beilstein 635639
Water 0.05% max.
Solubility Information Miscible in Water at 20°C
Formula Weight 60.11
Percent Purity >99.9%
Grade LC-MS
Chemical Name or Material Isopropyl Alcohol, B&J Brand™ LC-MS, for LC-MS and HPLC, >99.9%
Show More Show Less
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol