missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, B&J Brand™, for HPLC, GC, pesticide residue analysis, and spectrophotometry, >99.9%, Honeywell Burdick & Jackson
Supplier: Honeywell Chemicals 3234
Specifications
| Isopropyl Alcohol | |
| 67-63-0 | |
| 100.0000% | |
| 82.3°C | |
| 12°C | |
| C3H8O | |
| MFCD00008765 | |
| 635639 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| 60.11 | |
| HPLC |
| B&J Brand™, for HPLC, GC, pesticide residue analysis, and spectrophotometry, >99.9% | |
| 0.16 AU max. at 225 nm, 0.02 AU max. at 250 nm, 0.005 AU max. at 300 nm, 0.005 AU max. at 400 nm | |
| 0.785 g/cm3 | |
| 4 x 4 L | |
| Glass Bottle | |
| CH3COCH3 | |
| UN1219 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CC(C)O | |
| 60.096 | |
| CHEBI:17824 | |
| >99.9% |
Chemical Identifiers
| 67-63-0 | |
| 60.096 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
| C3H8O | |
| MFCD00008765 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol