missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%, Honeywell
Supplier: Honeywell Chemicals LP3235G
Specifications
| Isopropyl Alcohol | |
| 67-63-0 | |
| 0.785 g/cm3 | |
| 5 gal. | |
| Fiberboard Box | |
| C3H8O | |
| Passes test | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| 60.11 | |
| ACS Reagent |
| ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5% | |
| 100.0000% | |
| 82.3°C | |
| 12°C | |
| by GC Analysis | |
| UN1219 | |
| 635639 | |
| Miscible in Water at 20°C | |
| CC(C)O | |
| 60.096 | |
| CHEBI:17824 | |
| >99.5% |
Chemical Identifiers
| 67-63-0 | |
| 60.096 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
| C3H8O | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol