missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%, Solstice
Specifications
Specifications
| CAS Max % | 100.0000% |
| Density | 0.785 g/cm3 |
| Quantity | 204 L |
| Flash Point | 12°C |
| Packaging | Metal Drum |
| Water | Passes test |
| UN Number | UN1219 |
| Beilstein | 635639 |
| Boiling Point | 82.3°C |
| Solubility Information | Miscible in Water at 20°C |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol |