missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, >99.9%, Solstice

Catalog No. 6000879
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
Poly-coated Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000879 4 x 4 L Glass Bottle
6000880 4 x 4 L Poly-coated Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000879 Supplier Honeywell Inc Supplier No. AH3234

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Absorbance 0.16 AU max. at 225 nm, 0.02 AU max. at 250 nm
CAS Max % 100.0000%
Density 0.785 g/cm3
Quantity 4 x 4 L
Flash Point 12°C
Packaging Glass Bottle
Water 0.06% max.
UN Number UN1219
Beilstein 635639
Boiling Point 82.3°C
Solubility Information Miscible in Water at 20°C
Formula Weight 60.11
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Isopropyl Alcohol, ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol
EINECSNumber 200-661-7