missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, 99% (Molecular Biology Reagent), MP Biomedicals

Catalog No. ICN19400625
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19400625 25 mL
ICN19400680 100 mL
ICN19400690 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19400625 Supplier MP Biomedicals Inc Supplier No. 0219400625

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Quantity 25 mL
Formula Weight 60.09
Chemical Name or Material Isopropyl Alcohol, 99%