missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, 99% min., MilliporeSigma™
$6736.08 - $6736.08
Chemical Identifiers
| CAS | 67-63-0 |
|---|---|
| Molecular Formula | C3H8O |
| Molecular Weight (g/mol) | 60.096 |
| InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
| Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
| PubChem CID | 3776 |
| ChEBI | CHEBI:17824 |
| IUPAC Name | propan-2-ol |
| SMILES | CC(C)O |
Chemical Identifiers
| 67-63-0 | |
| 60.096 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
| C3H8O | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
Specifications
| 67-63-0 | |
| Metal Drum | |
| 99%, min. | |
| CH3CHOHCH3 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| 6.1 |
| 200 L | |
| Liquid | |
| C3H8O | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CC(C)O | |
| 60.096 | |
| CHEBI:17824 | |
| Isopropyl Alcohol |
Safety and Handling
Recommended Storage : Room Temperature