missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, 99%, EP, BP, JP, USP, 99%, Spectrum™ Chemical

Catalog No. 18603529
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603529 4 x 4 L Amber Glass Bottle
18603525 500 mL Amber Glass Bottle
18603532 1 L Amber Glass Bottle
18603526 6 x 500 mL Amber Glass Bottle
18603527 4 L Amber Glass Bottle
18603528 4 L Poly Bottle
18603533 6 x 1 L Amber Glass Bottle
18603530 4 x 4 L Poly Bottle
18603531 20 L Steel Can
18603534 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 18603529 Supplier Spectrum Chemical Mfg Cor Supplier No. IS1154LTGLCS4
Only null left

IS115, 67-63-0, C3H8O

Spectrum™ Chemical Isopropyl Alcohol, 99 Percent, USP, EP, BP, JP has many uses, including as a pharmaceutical raw material for preparing USP Rubbing Alcohol for relief of muscle stiffness and an infection preventative for minor scrapes and burns. All Spectrum Chemical USP, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 100%
Appearance Clear
Quantity 4 x 4 L
Packaging Amber Glass Bottle
Refractive Index 1.376-1.378
Specific Gravity 0.785-0.788
UN Number UN1219
Water 0.5%
Boiling Point 78.3°C
Percent Purity 99%
Grade Multi-Compendial
Chemical Name or Material Isopropyl Alcohol
Show More Show Less