missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, 99.0%, MilliporeSigma™

Catalog No. 50947587
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50947587 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50947587 Supplier Technidata Supplier No. PX18303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-63-0
Molecular Weight (g/mol) 60.1

Specifications

Melting Point −89.5°C
Quantity 20 L
Flash Point 12°C
Packaging Steel Drum
Residue after Evaporation 0.001% max
Assay Percent Range 99% min
Vapor Pressure 43 hPa (20°C)
Boiling Point 82.4°C (1013 hPa)
Recommended Storage Store at room temperature