missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, 70%, Reagent grade, MilliporeSigma™

Catalog No. MPX18403
Change view
Click to view available options
Quantity:
1 L
20 L
Packaging:
Cubitainer
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MPX18403 20 L Cubitainer
MPX18401 1 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MPX18403 Supplier Technidata Supplier No. PX18403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
Quantity 20 L
Packaging Cubitainer
Physical Form Liquid
Assay Percent Range 70%
Chemical Name or Material Isopropyl Alcohol