missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, 50% v/v, Reagents
Isopropyl Alcohol, 50% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CI2963004A
Specifications
| Isopropyl Alcohol, 50% v/v | |
| Colorless | |
| 4 L | |
| Liquid | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 50% |
| 7732-18-5,67-63-0 | |
| 81°C | |
| Poly Bottle | |
| 0.9257 | |
| Isopropyl Alcohol, 2-Propanol, Propan-2-ol | |
| O | |
| 18.02 | |
| Reagent |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Isopropyl Alcohol, 2-Propanol, Propan-2-ol | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |