missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Acetate 99.0+%, TCI America™

Supplier:  TCI America A003625ML

 View more versions of this product

Catalog No. A003625ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Isopropyl Acetate
Colorless
C5H10O2
25 mL
isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
CC(C)OC(=O)C
102.133
102.13
Liquid
108-21-4
88°C
MFCD00008877
1220
JMMWKPVZQRWMSS-UHFFFAOYSA-N
propan-2-yl acetate
7915
≥99.0% (GC)

Chemical Identifiers

108-21-4
102.133
JMMWKPVZQRWMSS-UHFFFAOYSA-N
7915
CC(C)OC(=O)C
C5H10O2
MFCD00008877
isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
propan-2-yl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0727

RTECSNumber : AI4930000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions