missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl 4-Hydroxybenzoate 99.0+%, TCI America™
Supplier: TCI America H0494500G
Specifications
| Isopropyl 4-Hydroxybenzoate | |
| 86°C | |
| 160°C | |
| MFCD00016468 | |
| isopropyl 4-hydroxybenzoate, isopropylparaben, isopropyl p-hydroxybenzoate, isopropyl paraben, p-hydroxybenzoic acid isopropyl ester, isopropylhydroxybenzoate, unii-a6eox47qk0, p-oxybenzoesaureisopropylester, 4-hydroxybenzoic acid isopropyl ester, benzoic acid, 4-hydroxy-, 1-methylethyl ester | |
| CC(C)OC(=O)C1=CC=C(O)C=C1 | |
| 180.20 | |
| 180.20 | |
| Powder or Flakes |
| 4191-73-5 | |
| White | |
| C10H12O3 | |
| 500 g | |
| CMHMMKSPYOOVGI-UHFFFAOYSA-N | |
| propan-2-yl 4-hydroxybenzoate | |
| 20161 | |
| ≥99.0% (T) |
Chemical Identifiers
| 4191-73-5 | |
| 180.20 | |
| CMHMMKSPYOOVGI-UHFFFAOYSA-N | |
| 20161 | |
| CC(C)OC(=O)C1=CC=C(O)C=C1 |
| C10H12O3 | |
| MFCD00016468 | |
| isopropyl 4-hydroxybenzoate, isopropylparaben, isopropyl p-hydroxybenzoate, isopropyl paraben, p-hydroxybenzoic acid isopropyl ester, isopropylhydroxybenzoate, unii-a6eox47qk0, p-oxybenzoesaureisopropylester, 4-hydroxybenzoic acid isopropyl ester, benzoic acid, 4-hydroxy-, 1-methylethyl ester | |
| propan-2-yl 4-hydroxybenzoate |
Safety and Handling
EINECSNumber : (3)-1585
RTECSNumber : DH2250000
TSCA : No