missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 4-Hydroxybenzoate 99.0+%, TCI America™

Catalog No. H049425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H049425G 25 g
H0494100G 100 g
H0494500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H049425G Supplier TCI America Supplier No. H049425G

Chemical Identifiers

CAS 4191-73-5
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00016468
InChI Key CMHMMKSPYOOVGI-UHFFFAOYSA-N
Synonym isopropyl 4-hydroxybenzoate, isopropylparaben, isopropyl p-hydroxybenzoate, isopropyl paraben, p-hydroxybenzoic acid isopropyl ester, isopropylhydroxybenzoate, unii-a6eox47qk0, p-oxybenzoesaureisopropylester, 4-hydroxybenzoic acid isopropyl ester, benzoic acid, 4-hydroxy-, 1-methylethyl ester
PubChem CID 20161
IUPAC Name propan-2-yl 4-hydroxybenzoate
SMILES CC(C)OC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 86°C
Color White
Boiling Point 160°C
Quantity 25 g
Formula Weight 180.20
Percent Purity ≥99.0% (T)
Physical Form Powder or Flakes
Chemical Name or Material Isopropyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
RTECSNumber DH2250000
TSCA No