Learn More
Isopropyl 4-hydroxybenzoate, 98%
CAS: 4191-73-5 | C10H12O3 | 180.20 g/mol
Supplier: Thermo Scientific Chemicals A1393018
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
ApplicationsIt is used as a preservative in cosmetics.
Solubility
Soluble in Chloroform, Ethyl Acetate.
Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
Specifications
| Isopropyl 4-hydroxybenzoate | |
| 84°C to 86°C | |
| MFCD00016468 | |
| 2641564 | |
| Soluble in Chloroform,Ethyl Acetate. | |
| CC(C)OC(=O)C1=CC=C(O)C=C1 | |
| 180.20 | |
| 180.21 |
| 4191-73-5 | |
| C10H12O3 | |
| 50 g | |
| isopropyl 4-hydroxybenzoate, isopropylparaben, isopropyl p-hydroxybenzoate, isopropyl paraben, p-hydroxybenzoic acid isopropyl ester, isopropylhydroxybenzoate, unii-a6eox47qk0, p-oxybenzoesaureisopropylester, 4-hydroxybenzoic acid isopropyl ester, benzoic acid, 4-hydroxy-, 1-methylethyl ester | |
| CMHMMKSPYOOVGI-UHFFFAOYSA-N | |
| propan-2-yl 4-hydroxybenzoate | |
| 20161 | |
| 98% |
Chemical Identifiers
| 4191-73-5 | |
| 180.20 | |
| CMHMMKSPYOOVGI-UHFFFAOYSA-N | |
| 20161 | |
| CC(C)OC(=O)C1=CC=C(O)C=C1 |
| C10H12O3 | |
| MFCD00016468 | |
| isopropyl 4-hydroxybenzoate, isopropylparaben, isopropyl p-hydroxybenzoate, isopropyl paraben, p-hydroxybenzoic acid isopropyl ester, isopropylhydroxybenzoate, unii-a6eox47qk0, p-oxybenzoesaureisopropylester, 4-hydroxybenzoic acid isopropyl ester, benzoic acid, 4-hydroxy-, 1-methylethyl ester | |
| propan-2-yl 4-hydroxybenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
H315-H317-H319-H335
EINECSNumber : 224-069-3
RTECSNumber : DH2250000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only