missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 4-hydroxybenzoate, 98%

Catalog No. AAA1393030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1393030 250 g
AAA1393009 10 g
AAA1393018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1393030 Supplier Thermo Scientific Chemicals Supplier No. A1393030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4191-73-5 | C10H12O3 | 180.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It is used as a preservative in cosmetics.

Solubility
Soluble in Chloroform, Ethyl Acetate.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.

Chemical Identifiers

CAS 4191-73-5
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00016468
InChI Key CMHMMKSPYOOVGI-UHFFFAOYSA-N
Synonym isopropyl 4-hydroxybenzoate, isopropylparaben, isopropyl p-hydroxybenzoate, isopropyl paraben, p-hydroxybenzoic acid isopropyl ester, isopropylhydroxybenzoate, unii-a6eox47qk0, p-oxybenzoesaureisopropylester, 4-hydroxybenzoic acid isopropyl ester, benzoic acid, 4-hydroxy-, 1-methylethyl ester
PubChem CID 20161
IUPAC Name propan-2-yl 4-hydroxybenzoate
SMILES CC(C)OC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 84°C to 86°C
Quantity 250 g
Beilstein 2641564
Solubility Information Soluble in Chloroform,Ethyl Acetate.
Formula Weight 180.21
Percent Purity 98%
Chemical Name or Material Isopropyl 4-hydroxybenzoate
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 224-069-3
RTECSNumber DH2250000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only