missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 4-aminobenzoate, 98%

Catalog No. p-7023720
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1755114 25 g
AAA1755122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1755114 Supplier Thermo Scientific Chemicals Supplier No. A1755114

CAS: 18144-43-9 | C10H13NO2 | 179.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18144-43-9
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00017108
InChI Key JWCPZKNBPMSYND-UHFFFAOYSA-N
Synonym isopropyl 4-aminobenzoate, 4-amino-benzoic acid isopropyl ester, isopropyl p-aminobenzoate, 4-aminobenzoic acid isopropyl ester, benzoic acid, 4-amino-, 1-methylethyl ester, benzoic acid, p-amino-, isopropyl ester, methylethyl 4-aminobenzoate, isopropyl-4-aminobenzoate, 4-isopropoxycarbonylaniline, acmc-1brt8
PubChem CID 87470
IUPAC Name propan-2-yl 4-aminobenzoate
SMILES CC(C)OC(=O)C1=CC=C(C=C1)N

Specifications

Quantity 25 g
Formula Weight 179.21
Percent Purity 98%
Chemical Name or Material Isopropyl 4-aminobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only