missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 4,4,4-Trifluoroacetoacetate 95.0+%, TCI America™

Catalog No. t153025g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T153025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T153025G Supplier TCI America Supplier No. T153025G

Chemical Identifiers

CAS 175230-50-9
Molecular Formula C7H9F3O3
Molecular Weight (g/mol) 198.14
MDL Number MFCD00040990
InChI Key XLBGYZLICFMDDS-UHFFFAOYSA-N
Synonym isopropyl 4,4,4-trifluoroacetoacetate, isopropyl 4,4,4-trifluoro-3-oxobutanoate, isopropyl 4,4,4-trifluoro-3-oxobutyrate, isopropyl-4,4,4-trifluoroacetoacetate, acmc-209e9i, isopropyl4,4,4-trifluoroacetoacetate, methylethyl 4,4,4-trifluoro-3-oxobutanoate, isopropyl 4,4,4-trifluoro-3-oxobutanoate #, 4,4,4-trifluoroacetoacetic acid isopropyl ester
PubChem CID 586897
IUPAC Name propan-2-yl 4,4,4-trifluoro-3-oxobutanoate
SMILES CC(C)OC(=O)CC(=O)C(F)(F)F

Specifications

Color Colorless
Quantity 25 g
UN Number 3272
Formula Weight 198.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl 4,4,4-Trifluoroacetoacetate