missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 3-Aminocrotonate 98.0+%, TCI America™

Catalog No. p-7131641
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08841G 1 g
I08845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08841G Supplier TCI America Supplier No. I08841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14205-46-0
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD00134272
InChI Key YCKAGGHNUHZKCL-XQRVVYSFSA-N
Synonym 3-Aminocrotonic Acid Isopropyl Ester, Isopropyl 3-Amino-2-butenoate, 3-Amino-2-butenoic Acid Isopropyl Ester
PubChem CID 11137274
IUPAC Name propan-2-yl (Z)-3-aminobut-2-enoate
SMILES CC(C)OC(=O)C=C(C)N

Specifications

Color Yellow
Boiling Point 80°C
Quantity 1 g
Formula Weight 143.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Isopropyl 3-Aminocrotonate
RTECSNumber EM9095000
TSCA No