accessibility menu, dialog, popup

UserName

CAS RN 14205-46-0

IUPAC Name: propan-2-yl 3-aminobut-2-enoate
Synonyms: isopropyl 3-aminobut-2-enoate
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: YCKAGGHNUHZKCL-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)C=C(C)N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results