missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropanol/Water, 91:9, Reagents
Isopropanol/Water, 91:9, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CI298100500A
Specifications
| Isopropanol/Water, 91:9 | |
| Colorless | |
| 500 mL | |
| Liquid | |
| C3H8O | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 91% |
| 67-63-0,7732-18-5 | |
| 82°C | |
| Poly Bottle | |
| 0.8159 | |
| Isopropyl Alcohol, 2-Propanol, Propan-2-ol | |
| CC(C)O | |
| 60.10 | |
| Reagent |
Chemical Identifiers
| 67-63-0 | |
| 60.10 | |
| Isopropyl Alcohol, 2-Propanol, Propan-2-ol | |
| CC(C)O |
| C3H8O | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol |