missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, Molecular Biology Grade, Fisher BioReagents™ Edge
Edge

For nucleic acid precipitation | CAS: 67-63-0 | C3H8O | 60.096 g/mol

Supplier:  Fisher BioReagents BP26181

 Afficher plus de versions de ce produit

Numéro de catalogue. BP26181



Il en reste null
Edge
Découvrez plus d'offres spéciales

Description

Description

For nucleic acid precipitation

Ultrapure reagent that can be used in fundamental applications such as purification and precipitation of nucleic acids.
Spécifications

Spécifications

7
-89.5°C
0.015 AU max. at 254nm, 0.10 AU max. at 230nm, 0.20 AU max. at 220nm, 1.00 AU max. at 205nm
0.785g/cm³
1 L
Liquid
1.3740 to 1.3760 (at 25°C)
≥99.9 %
(CH3)2CHOH
2.27 mPa/s at 20°C
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
60.096
CHEBI:17824
≥99.9%
Isopropanol
67-63-0
Colorless
81°C
Amber Glass Bottle
DNase-, RNase- and Protease-Free
1ppm max.
C3H8O
43mmHg at 20°C
0.05% max.
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1
Molecular Biology

Identifiants chimiques

67-63-0
60.096
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

WARNING!

Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0

missing translation for 'dotInformation' : DOT Class 3, : Flammable Liquid

FDS Documents

Documents

Promotions

Promotions