missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, Molecular Biology Grade, Fisher BioReagents™

Catalog No. BP26184
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP26184 4 L
BP2618500 500 mL
BP26181 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP26184 Supplier Fisher BioReagents Supplier No. BP2618 4
Edge

For nucleic acid precipitation | CAS: 67-63-0 | C3H8O | 60.096 g/mol

For nucleic acid precipitation

Ultrapure reagent that can be used in fundamental applications such as purification and precipitation of nucleic acids.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH 7
Melting Point -89.5°C
Absorbance 0.015 AU max. at 254nm, 0.10 AU max. at 230nm, 0.20 AU max. at 220nm, 1.00 AU max. at 205nm
Color Colorless
Density 0.785g/cm³
Quantity 4 L
Packaging Amber Glass Bottle
Physical Form Liquid
Refractive Index 1.3740 to 1.3760 (at 25°C)
Residue after Evaporation 1ppm max.
Assay Percent Range ≥99.9 %
Vapor Pressure 43mmHg at 20°C
Linear Formula (CH3)2CHOH
Viscosity 2.27 mPa/s at 20°C
Water 0.05% max.
Boiling Point 81°C
Formula Weight 60.1
Percent Purity ≥99.9%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Isopropanol
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid