missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, ACS Reagent Grade, Ricca Chemical

Catalog No. p-3340394
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RSOI00201C 1 L Amber Glass Bottle
RSOI00204C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RSOI00201C Supplier Ricca Chemical Company Supplier No. RSOI00201C
Only null left

For General Use

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
CAS Min % 98.0
CAS Max % 100.0
Boiling Point 82.5°C
Quantity 1 L
Packaging Amber Glass Bottle
Physical Form Liquid
Residue after Evaporation max. 0.001%
Water max. 0.2%
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Molecular Weight (g/mol) 60.096
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Isopropanol
Show More Show Less
ShelfLife 24 months