missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropanol, ACS Reagent (2-Propanol; Isopropyl Alcohol), Reagents
Specifications
Specifications
| Melting Point | -90°C |
| Color | Colorless |
| Boiling Point | 82.5°C |
| Quantity | 10 L |
| Packaging | Steel Pail |
| Physical Form | Liquid |
| Specific Gravity | 0.7808 |
| Grade | ACS |
| Chemical Name or Material | Isopropanol, ACS Reagent (2-Propanol, Isopropyl Alcohol) |