missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, ACS Reagent (2-Propanol; Isopropyl Alcohol), Reagents

Catalog No. 01338124
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
200 L
Packaging:
Cubitainer
Glass Bottle
ISOPROPANOL (IPA) ACS
Poly Bottle
Safety Coated Glass
Steel Pail
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01338124 10 L Steel Pail
01338123 1 L Glass Bottle
01338122 200 L ISOPROPANOL (IPA) ACS
01338121 20 L Steel Pail
01338120 20 L Cubitainer
01338119 4 L Poly Bottle
01338118 4 L Glass Bottle
01338117 4 L Safety Coated Glass
01338116 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 01338124 Supplier Labchem Inc Supplier No. C112150010E6

Isopropanol, ACS Reagent (2-Propanol; Isopropyl Alcohol), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym Isopropyl Alcohol, 2-Propanol, Propan-2-ol
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -90°C
Color Colorless
Quantity 10 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.7808
Boiling Point 82.5°C
Grade ACS
Chemical Name or Material Isopropanol, ACS Reagent (2-Propanol, Isopropyl Alcohol)