missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, 99.5%, for HPLC

CAS: 67-63-0 | C3H8O | 60.096 g/mol

$141.53 - $141.53

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC383910010
SDS View Documents
Thermo Scientific Chemicals
383910010
1 L
Each for $141.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67-63-0
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
C3H8O
MFCD00011674
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O

Specifications

-89.5°C
0.7850g/mL
1 L
Glass Bottle
99.5%
C3H8O
MFCD00011674
15, 5254
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride,, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions
CC(C)O
60.096
CHEBI:17824
99.5%
Isopropanol
67-63-0
81.0°C to 83.0°C
12°C
1.3760 to 1.3780
0.785
(CH3)2CHOH
2.27 mPa.s (20°C)
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1
HPLC
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-661-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only