missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, 70% (v/v), Ricca Chemical
Supplier: Ricca Chemical Company 421055
Specifications
| Isopropanol | |
| 29.0,69.0 | |
| Colorless Liquid | |
| 55 gal. | |
| 0.874 to 0.882 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CC(C)O | |
| Mixture | |
| CHEBI:17824 | |
| Laboratory |
| 67-63-0,7732-18-5 | |
| 31.0,71.0 | |
| 82°C | |
| Poly Drum | |
| C3H8O | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| 68 to 72% v/v |
Chemical Identifiers
| 67-63-0 | |
| Mixture | |
| 3776 | |
| propan-2-ol |
| C3H8O | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| CHEBI:17824 | |
| CC(C)O |
Safety and Handling
ShelfLife : 24 months