missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, 70% (v/v), Ricca Chemical

Supplier:  Ricca Chemical Company RI42101

 View more versions of this product

Catalog No. 4210-1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Isopropanol
29.0,69.0
Colorless Liquid
4 L
0.874 to 0.882
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
Mixture
CHEBI:17824
Laboratory
67-63-0,7732-18-5
31.0,71.0
82°C
Natural Poly Bottle
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
68 to 72% v/v

Chemical Identifiers

67-63-0
Mixture
3776
propan-2-ol
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
CHEBI:17824
CC(C)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS Documents

Documents

Promotions

Promotions