missing translation for 'onlineSavingsMsg'
Learn More

Isoprene, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7193412
Change view
Click to view available options
Quantity:
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020325 1 mL
111018312 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020325 Supplier Merck Emd Millipore Supplier No. 592401MLF
Only null left

Isoprene is a reactive aliphatic hydrocarbon that originates as a byproduct of cholesterol synthesis.

Specifications

CAS 78-79-5
Solution Contains ˜0.02% 4-tert-butylcatechol as stabilizer
Vapor Pressure 8.82 psi (20°C)
Vapor Density 2.35 (vs. air)
Density 0.681 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH2=CHC(CH3)=CH2
MDL Number MFCD00008600
Melting Point -146°C
Boiling Point 34°C
Synonym 2-Methyl-1,3-butadiene
Quantity 1 mL
Formula Weight 68.12
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less