missing translation for 'onlineSavingsMsg'
Learn More

Isophthalic Acid 99.0+%, TCI America™

Catalog No. p-7131239
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I015525G 25 g
I0155500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I015525G Supplier TCI America Supplier No. I015525G
Only null left

Chemical Identifiers

CAS 121-91-5
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.132
MDL Number MFCD00002516
InChI Key QQVIHTHCMHWDBS-UHFFFAOYSA-N
Synonym isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj
PubChem CID 8496
ChEBI CHEBI:30802
IUPAC Name benzene-1,3-dicarboxylic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 340°C
Color White
Quantity 25 g
Formula Weight 166.13
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isophthalic Acid
EINECSNumber (3)-1332
RTECSNumber NT2007000
TSCA Yes