accessibility menu, dialog, popup

UserName

CAS RN 121-91-5

IUPAC Name: benzene-1,3-dicarboxylic acid
Synonyms: isophthalic acid kyselina isoftalova m-phthalic acid acide isophtalique
Molecular Weight (g/mol): 166.13
Molecular Formula: C8H6O4
InChi Key: QQVIHTHCMHWDBS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)