missing translation for 'onlineSavingsMsg'
Learn More

Isophthalic acid, 99%

CAS: 121-91-5 | C8H6O4 | 166.132 g/mol

Supplier:  Thermo Scientific Chemicals A144450I

 View more versions of this product

Catalog No. AAA144450I

Only null left
Explore available promotions

Description

Description

Isophthalic acid is used as an intermediate for high performance unsaturated polyesters, resins for coatings, high solids paints, gel coats and modifier of polyethylene terephthalate for bottles. It acts as precursors for the fire-resistant material nomex as well as used in the preparation of high-performance polymer polybenzimidazole. It is also employed as an input for the production of insulation materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophthalic acid is used as an intermediate for high performance unsaturated polyesters, resins for coatings, high solids paints, gel coats and modifier of polyethylene terephthalate for bottles. It acts as precursors for the fire-resistant material nomex as well as used in the preparation of high-performance polymer polybenzimidazole. It is also employed as an input for the production of insulation materials.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

Isophthalic acid
344°C to 348°C (sublimation)
C8H6O4
5000 g
14,5197
Insoluble in water.
C1=CC(=CC(=C1)C(=O)O)C(=O)O
166.132
CHEBI:30802
99%
121-91-5
1.54
MFCD00002516
1909332
isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj
QQVIHTHCMHWDBS-UHFFFAOYSA-N
benzene-1,3-dicarboxylic acid
8496
166.13

Chemical Identifiers

121-91-5
166.132
QQVIHTHCMHWDBS-UHFFFAOYSA-N
8496
benzene-1,3-dicarboxylic acid
C8H6O4
MFCD00002516
isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj
CHEBI:30802
C1=CC(=CC(=C1)C(=O)O)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-506-4

RTECSNumber : NT2007000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only