Learn More
Isophthalic acid, 99%
CAS: 121-91-5 | C8H6O4 | 166.132 g/mol
Supplier: Thermo Scientific Chemicals A144450B
Description
Isophthalic acid is used as an intermediate for high performance unsaturated polyesters, resins for coatings, high solids paints, gel coats and modifier of polyethylene terephthalate for bottles. It acts as precursors for the fire-resistant material nomex as well as used in the preparation of high-performance polymer polybenzimidazole. It is also employed as an input for the production of insulation materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsophthalic acid is used as an intermediate for high performance unsaturated polyesters, resins for coatings, high solids paints, gel coats and modifier of polyethylene terephthalate for bottles. It acts as precursors for the fire-resistant material nomex as well as used in the preparation of high-performance polymer polybenzimidazole. It is also employed as an input for the production of insulation materials.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
| Isophthalic acid | |
| 344°C to 348°C (sublimation) | |
| C8H6O4 | |
| 1000 g | |
| 14,5197 | |
| Insoluble in water. | |
| C1=CC(=CC(=C1)C(=O)O)C(=O)O | |
| 166.132 | |
| CHEBI:30802 | |
| 99% |
| 121-91-5 | |
| 1.54 | |
| MFCD00002516 | |
| 1909332 | |
| isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj | |
| QQVIHTHCMHWDBS-UHFFFAOYSA-N | |
| benzene-1,3-dicarboxylic acid | |
| 8496 | |
| 166.13 |
Chemical Identifiers
| 121-91-5 | |
| 166.132 | |
| QQVIHTHCMHWDBS-UHFFFAOYSA-N | |
| 8496 | |
| benzene-1,3-dicarboxylic acid |
| C8H6O4 | |
| MFCD00002516 | |
| isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj | |
| CHEBI:30802 | |
| C1=CC(=CC(=C1)C(=O)O)C(=O)O |
Safety and Handling
EINECSNumber : 204-506-4
RTECSNumber : NT2007000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only