missing translation for 'onlineSavingsMsg'
Learn More

Isophorone Oxide 97.0+%, TCI America™

Catalog No. i08055g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08055G 5 g
I080525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08055G Supplier TCI America Supplier No. I08055G
Only null left

Chemical Identifiers

CAS 10276-21-8
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
MDL Number MFCD00085465
InChI Key ROTNQFPVXVZSRL-UHFFFAOYSA-N
Synonym 3,5,5-Trimethyl-2,3-epoxy-1-cyclohexanone, 2,3-Epoxy-3,5,5-trimethyl-1-cyclohexanone, 4,4,6-Trimethyl-7-oxabicyclo[4.1.0]heptan-2-one
PubChem CID 25121
IUPAC Name 4,4,6-trimethyl-7-oxabicyclo[4.1.0]heptan-2-one
SMILES CC1(CC(=O)C2C(C1)(O2)C)C

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 g
Formula Weight 154.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isophorone Oxide