accessibility menu, dialog, popup

UserName

CAS RN 10276-21-8

IUPAC Name: 4,4,6-trimethyl-7-oxabicyclo[4.1.0]heptan-2-one
Synonyms: 4,4,6-trimethyl-7-oxabicyclo[4.1.0]heptan-2-one
Molecular Weight (g/mol): 154.21
Molecular Formula: C9H14O2
InChi Key: ROTNQFPVXVZSRL-UHFFFAOYNA-N
SMILES: CC12CC(C)(C)CC(=O)C1O2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results