missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isophorone Diisocyanate (mixture of isomers) 99.0+%, TCI America™
Supplier: TCI America I0314500ML
Specifications
| Isophorone Diisocyanate (mixture of isomers) | |
| -60°C | |
| 159°C | |
| MFCD00064956 | |
| 2290 | |
| NIMLQBUJDJZYEJ-UHFFFAOYNA-N | |
| 5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane | |
| 169132 | |
| 222.29 | |
| Liquid |
| 4098-71-9 | |
| Colorless | |
| C12H18N2O2 | |
| 500 mL | |
| isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates | |
| CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O | |
| 222.29 | |
| CHEBI:53214 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 4098-71-9 | |
| 222.29 | |
| NIMLQBUJDJZYEJ-UHFFFAOYNA-N | |
| 169132 | |
| 5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane |
| C12H18N2O2 | |
| MFCD00064956 | |
| isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates | |
| CHEBI:53214 | |
| CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O |
Safety and Handling
EINECSNumber : (3)-2492
RTECSNumber : NQ9370000
TSCA : Yes