missing translation for 'onlineSavingsMsg'
Learn More

Isophorone Diisocyanate (mixture of isomers) 99.0+%, TCI America™

Catalog No. p-7131327
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I031425ML 25 mL
I0314500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I031425ML Supplier TCI America Supplier No. I031425ML
Only null left

Chemical Identifiers

CAS 4098-71-9
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.29
MDL Number MFCD00064956
InChI Key NIMLQBUJDJZYEJ-UHFFFAOYNA-N
Synonym isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates
PubChem CID 169132
ChEBI CHEBI:53214
IUPAC Name 5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
SMILES CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O

Specifications

Melting Point -60°C
Color Colorless
Boiling Point 159°C
Quantity 25 mL
UN Number 2290
Formula Weight 222.29
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isophorone Diisocyanate (mixture of isomers)
EINECSNumber (3)-2492
RTECSNumber NQ9370000
TSCA Yes