missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isooctane, Uvasol™, MilliporeSigma™
For spectroscopy Uvasol™
$214.48 - $767.66
Chemical Identifiers
| CAS | 540-84-1 |
|---|---|
| Molecular Formula | C8H18 |
| Molecular Weight (g/mol) | 114.232 |
| InChI Key | NHTMVDHEPJAVLT-UHFFFAOYSA-N |
| Synonym | isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane |
| PubChem CID | 10907 |
| ChEBI | CHEBI:62805 |
| IUPAC Name | 2,2,4-trimethylpentane |
| SMILES | CC(C)CC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1047180500
|
Technidata
1047180500 |
500 mL | Glass bottle |
Each for $214.48
|
|
||||
|
M1047182500
|
Technidata
1047182500 |
2.5 L | Glass bottle |
Each for $767.66
|
|
||||
Chemical Identifiers
| 540-84-1 | |
| 114.232 | |
| isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
| CHEBI:62805 | |
| CC(C)CC(C)(C)C |
| C8H18 | |
| NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
| 10907 | |
| 2,2,4-trimethylpentane |
Specifications
| 540-84-1 | |
| Colorless | |
| 99°C | |
| 410°C (770°F) | |
| C8H18 | |
| isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
| NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
| 2,2,4-trimethylpentane | |
| 10907 | |
| 114.23 | |
| Liquid |
| -107°C | |
| 0.69g/cm3 (20°C) | |
| -12°C (+10°F) | |
| Glass bottle | |
| 500 mL | |
| Water: 0.56mg/L at 25°C soluble | |
| CC(C)CC(C)(C)C | |
| 114.232 | |
| CHEBI:62805 | |
| 51 hPa (20°C) | |
| Isooctane |
Safety and Handling
H304: May be fatal if swallowed and enters airways.
H315: Causes skin irritation.
H225: Highly flammable liquid and vapor
DOTInformation : DOT Class: 3, Packaging Group: II
EINECSNumber : 208-759-1
RTECSNumber : SA3320000
Recommended Storage : Keep away from heat and sources of ignition. Keep container tightly closed in a dry and well-ventilated place.