missing translation for 'onlineSavingsMsg'
Learn More

Isooctane, SupraSolv™, MilliporeSigma™

Catalog No. M1154402500
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1154402500 2.5 L Glass bottle
M1154401000 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1154402500 Supplier Technidata Supplier No. 1.15440.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For gas chromatography ECD and FID SupraSolv™

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Melting Point -107°C
Color Colorless
Density 0.69g/cm3 (20°C)
Boiling Point 99°C
Flash Point -12°C (+10°F)
Ignition Point 410°C (770°F)
Packaging Glass bottle
Quantity 2.5 L
UN Number UN 1262
Solubility Information Water: 0.56mg/L at 25°C soluble
Vapor Pressure 51 hPa (20°C)
Formula Weight 114.23
Physical Form Liquid
Chemical Name or Material Isooctane
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapor
Hazard Statement H304: May be fatal if swallowed and enters airways.
Precautionary Statement H315: Causes skin irritation.
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 208-759-1
RTECSNumber SA3320000
Recommended Storage Keep away from heat and sources of ignition. Keep container tightly closed in a dry and well-ventilated place.