missing translation for 'onlineSavingsMsg'
Learn More

Isooctane (HPLC), Fisher Chemical™

Catalog No. p-216089
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Safe-Cote™ Amber Glass 
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
O296SK1 1 L Safe-Cote™ Amber Glass 
O2964 4 L Glass Bottle
O296SK4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O296SK1 Supplier Thermo Fisher Scientific Supplier No. O296SK1
Only null left

Submicron filtered

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Melting Point -107°C
Absorbance 0.014 AU max. at 254nm, 0.1 AU max. at 225nm, 1.0 AU max. at 205nm
Color Colorless
Density 0.690g/cm³
Quantity 1 L
Flash Point -12°C (10.4°F)
Identification Pass Test
Odor Petroleum distillates
Packaging Safe-Cote™ Amber Glass 
Physical Form Liquid
Refractive Index 1.3885 to 1.3905 (at 25°C)
Residue after Evaporation 4ppm max.
Assay Percent Range ≥99 %
Vapor Pressure 51mbar at 20°C
Viscosity 0.51 mPa/s at 22°C
UN Number 1262
Water 0.02% max.
Boiling Point 98°C
Solubility Information Solubility in water: insoluble
Formula Weight 114.23
Percent Purity ≥99%
Grade Also meets ACS specifications
Chemical Name or Material Isooctane
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA:

Health: 3
Flammability: 3
Instability: 0
DOTInformation DOT Class 3, : Flammable Liquid
Recommended Storage Keep away from open flames, hot surfaces and sources of ignition. Keep containers tightly closed in a dry, cool and well-ventilated place. Flammables area. Keep away from heat and sources of ignition.