missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinic anhydride, 90%, remainder picolinic and nicotinic acid-anhydride

Catalog No. AC250280050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC250280050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC250280050 Supplier Thermo Scientific Chemicals Supplier No. 250280050
Only null left

CAS: 7082-71-5 | C12H8N2O3 | 228.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7082-71-5
Molecular Formula C12H8N2O3
Molecular Weight (g/mol) 228.2
MDL Number MFCD00062784
InChI Key ORBIGZZHMAJXAZ-UHFFFAOYSA-N
Synonym isonicotinic anhydride, isonictinic anhydride, acmc-1bcxw, isonicotinic acid anhydride, bis 4-pyridinecarboxylic anhydride, 4-pyridinecarboxylicacid, anhydride with 4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid anhydride; pyridine-4-carboxylic acid anhydride
PubChem CID 81529
IUPAC Name pyridine-4-carbonyl pyridine-4-carboxylate
SMILES C1=CN=CC=C1C(=O)OC(=O)C2=CC=NC=C2

Specifications

Melting Point 133°C
Color Beige-Brown to White
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: 5g/L (20°C). Other solubilities: hydrolysis in water
Formula Weight 228.2
Percent Purity 90%
Physical Form Crystalline Powder
Chemical Name or Material Isonicotinic anhydride, 90%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-382-6

RUO – Research Use Only