accessibility menu, dialog, popup

UserName

OOONN

CAS RN 7082-71-5

IUPAC Name: pyridine-4-carbonyl pyridine-4-carboxylate
Synonyms: pyridine-4-carbonyl pyridine-4-carboxylate
Molecular Weight (g/mol): 228.21
Molecular Formula: C12H8N2O3
InChi Key: ORBIGZZHMAJXAZ-UHFFFAOYSA-N
SMILES: O=C(OC(=O)C1=CC=NC=C1)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results