missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isonicotinic Acid Hydrazide 98.0+%, TCI America™
Supplier: TCI America I0138100G
Specifications
| Isonicotinic Acid Hydrazide | |
| 172°C | |
| C6H7N3O | |
| 100 g | |
| QRXWMOHMRWLFEY-UHFFFAOYSA-N | |
| pyridine-4-carbohydrazide | |
| 3767 | |
| 137.14 | |
| Crystalline Powder |
| 54-85-3 | |
| White | |
| MFCD00006426 | |
| isoniazid, isonicotinic acid hydrazide, isonicotinohydrazide, rimifon, isoniazide, nydrazid, isonicotinic hydrazide, laniazid, isonicotinylhydrazine, andrazide | |
| C1=CN=CC=C1C(=O)NN | |
| 137.142 | |
| CHEBI:6030 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 54-85-3 | |
| 137.142 | |
| QRXWMOHMRWLFEY-UHFFFAOYSA-N | |
| 3767 | |
| pyridine-4-carbohydrazide |
| C6H7N3O | |
| MFCD00006426 | |
| isoniazid, isonicotinic acid hydrazide, isonicotinohydrazide, rimifon, isoniazide, nydrazid, isonicotinic hydrazide, laniazid, isonicotinylhydrazine, andrazide | |
| CHEBI:6030 | |
| C1=CN=CC=C1C(=O)NN |
Safety and Handling
EINECSNumber : (5)-0737
RTECSNumber : NS1751850
TSCA : Yes