missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinic Acid Hydrazide 98.0+%, TCI America™

Catalog No. I013825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I013825G 25 g
I0138100G 100 g
I0138500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I013825G Supplier TCI America Supplier No. I013825G

Chemical Identifiers

CAS 54-85-3
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.142
MDL Number MFCD00006426
InChI Key QRXWMOHMRWLFEY-UHFFFAOYSA-N
Synonym isoniazid, isonicotinic acid hydrazide, isonicotinohydrazide, rimifon, isoniazide, nydrazid, isonicotinic hydrazide, laniazid, isonicotinylhydrazine, andrazide
PubChem CID 3767
ChEBI CHEBI:6030
IUPAC Name pyridine-4-carbohydrazide
SMILES C1=CN=CC=C1C(=O)NN

Specifications

Melting Point 172°C
Color White
Quantity 25 g
Formula Weight 137.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isonicotinic Acid Hydrazide
EINECSNumber (5)-0737
RTECSNumber NS1751850
TSCA Yes